Merck
BAF182020000
SP Bel-Art® Chromatography Column Holder
diam. × H 12.25 in. (311 mm) × 2.5 in. (64 mm)
Synonym(e):
column holder
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.43
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F18202-0000
Durchm. × H
12.25 in. (311 mm) × 2.5 in. (64 mm)
Allgemeine Beschreibung
Securely holds as many as eight chromatography columns up to 30mm (1³⁄₁₆") in diameter in or between its flexible loops.
Leistungsmerkmale und Vorteile
  • Rigid plastic back is mounted on two polyethylene support blocks which slide over support stand rods
  • Tighten the screws to mount the rack at the desired height
  • Requires two support rods and stands (H18303-0000 sold separately) or can be wall-mounted by removing the support blocks from the back of the unit
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)