Skip to Content
Merck
All Photos(3)

Key Documents

PRS3877

Sigma-Aldrich

Anti-TANK (ab1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-I-TRAF, Anti-TRAF family member-associated NFKB activator

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 47 kDa

species reactivity

mouse, rat, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TANK(10010)

Immunogen

a 14 amino acid peptide from near the carboxy-terminus of human TANK.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3877.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai-ying Chen et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 68(6), 709-713 (2014-08-06)
Exposure of Tca-8113 cells to proteasome inhibitor carbobenzoxy-Leu-Leu-leucinal (MG-132) causing apoptosis is associated with endoplasmic reticulum (ER) stress. X-box-binding protein-1 (XBP1) is an important regulator of a subset of genes active during ER stress, which is related to cell survival
Juhee Jeon et al.
Biochemical pharmacology, 89(4), 515-525 (2014-04-17)
The ligation of interleukin-1 receptor (IL-1R) or tumor necrosis factor receptor 1 (TNFR1) induces the recruitment of adaptor proteins and their concomitant ubiquitination to the proximal receptor signaling complex, respectively. Such are upstream signaling events of IKK that play essential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service