03-191
RIPAb+ Upf1 - RIP Validated Antibody and Primer Set
from rabbit
Synonym(s):
ATP-dependent helicase RENT1, Nonsense mRNA reducing factor 1, UP Frameshift 1, UPF1 regulator of nonsense transcripts homolog (yeast), Up-frameshift suppressor 1 homolog, delta helicase, regulator of nonsense transcripts 1, up-frameshift mutation 1 homo
About This Item
Recommended Products
biological source
rabbit
Quality Level
clone
polyclonal
species reactivity
human
species reactivity (predicted by homology)
mouse, bovine, canine, rat
manufacturer/tradename
RIPAb+
Upstate®
technique(s)
RIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
General description
Specificity
Immunogen
Application
Representative lot data.
RIP lysate from HeLa cells (~2 X 10E7 cell equivalents per IP) was subjected to immunoprecipitation using either 7 µL of a normal rabbit IgG or 7 µL of Anti-Upf1 antibody.
Ten percent of the precipitated proteins (lane 1: rabbit IgG, lane 2: Upf1) were resolved by electrophoresis, transferred to PVDF and probed with anti-Upf1 antibody (1:2000).
Proteins were visualized using One-Step IP-Western kit (GenScript Cat. # L00232)
Arrow indicates Upf1 (~125 kDa) (Please see figures).
Western Blot Analysis:
Representative lot data.
U2OS cell lysate was resolved by electrophoresis, transferred to PVDF membrane and probed with anti-Upf1 (0.5 μg/mL).
Proteins were visualized using a donkey anti-rabbit secondary antibody conjugated to HRP and a chemiluminescence detection system.
Arrow indicates Upf1 (~125 kDa) (Please see figures).
Epigenetics & Nuclear Function
RNA Binding Protein (RBP)
Packaging
Quality
RIP Lysate prepared from HeLa cells (2 X 10E7 cell equivalents per IP) were subjected to immunoprecipitation using either 7 µL of a normal rabbit IgG or 7 µL Anti-Upf1 antibody and the Magna RIP® RNA-Binding Protein Immunoprecipitation Kit (Cat. # 17-700).
Successful immunoprecipitation of Upf1-associated RNA was verified by qPCR using RIP Primers ARF1 (Please see figures).
Please refer to the Magna RIP (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.
Target description
Physical form
Normal Rabbit IgG. One vial containing 125 µg of rabbit IgG in 125 µL of storage buffer containing 0.05% sodium azide. Store at -20°C.
RIP Primers, ARF1. One vial containing 75 μL of 5 μM of each primer specific for human ARF1.
Store at -20°C.
FOR: GAC CAC GAT CCT CTA CAA GC
REV: TCC CAC ACA GTG AAG CTG ATG
Storage and Stability
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.
Analysis Note
Includes negative control normal rabbit IgG antibody and control primers specific for the cDNA of human ARF1.
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service