Skip to Content
Merck
All Photos(3)

Key Documents

AV02062

Sigma-Aldrich

Anti-YWHAQ antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-1C5, Anti-HS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

28 kDa

species reactivity

human, mouse, rat, guinea pig, dog, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... YWHAQ(10971)

Immunogen

Synthetic peptide directed towards the N terminal region of human YWHAQ

Application

Anti-YWHAQ antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml. For immunohistochemistry of paraffin-embedded tissue sections, a concentration of 4-8 μg/ml is suitable.

Biochem/physiol Actions

YWHAQ is a regulator of intrinsic apoptotic pathway. Promoter variation of YWHAQ gene reduces cell death and affects the treatment outcomes in childhood acute lymphoblastic leukemia.

Sequence

Synthetic peptide located within the following region: SVAYKNVVGGRRSAWRVISSIEQKTDTSDKKLQLIKDYREKVESELRSIC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rocio Sanchez et al.
Haematologica, 99(2), 314-321 (2013-09-17)
The introduction of multiagent treatment protocols has led to a remarkable increase in survival rates for children diagnosed with acute lymphoblastic leukemia, yet for a subpopulation of patients, resistance to chemotherapeutics remains an obstacle to successful treatment. Here we investigate
M Iwamoto et al.
Free radical research, 48(12), 1409-1416 (2014-09-03)
We here described the antioxidant effects of carnitine supplementation on 14-3-3 protein isoforms in the aged rat hippocampus detected using the fully automated two-dimensional chip gel electrophoresis system (Auto2D). This system was easy and convenient to use, and the resolution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service