GF93927516
Inconel® alloy 600 - heat resisting alloy, Ni72Cr16Fe8
foil, 150 x 150mm, 0.0125mm thickness, annealed, not light tested (nlt)
Synonym(s):
NI020203
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101712
NACRES:
NA.23
description
not light tested (nlt)
form
annealed (
quarter hard)
foil (
quarter hard)
manufacturer/tradename
Goodfellow 939-275-16
size × thickness
150 mm × 150 mm × 0.0125 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service