Merck
  • Minimal active domain and mechanism of action of the angiogenesis inhibitor histidine-rich glycoprotein.
Minimal active domain and mechanism of action of the angiogenesis inhibitor histidine-rich glycoprotein.
Cancer research (2006-02-21)
Johan Dixelius, Anna-Karin Olsson, Asa Thulin, Chunsik Lee, Irja Johansson, Lena Claesson-Welsh
ABSTRACT
Histidine-rich glycoprotein (HRGP) is an abundant heparin-binding plasma protein that efficiently arrests growth and vascularization of mouse tumor models. We have shown that the antiangiogenic effect of HRGP is dependent on its histidine/proline-rich domain, which needs to be released from the mother protein to exert its effects. Here we identify a 35-amino-acid peptide, HRGP330, derived from the histidine/proline-rich domain as endowed with antiangiogenic properties in vitro and in vivo. The mechanism of action of HRGP330 involves subversion of focal adhesion function by disruption of integrin-linked kinase (ILK) and focal adhesion kinase (FAK) functions, inhibition of vascular endothelial growth factor (VEGF)-induced tyrosine phosphorylation of the FAK substrate alpha-actinin, and, as a consequence, an arrest in endothelial cell motility. The disturbed focal adhesion function is reflected in the ability of HRGP as well as of HRGP330 to prevent endothelial cell adhesion to vitronectin in a manner involving alpha(v)beta3 integrin. In conclusion, HRGP330, which we define as the minimal antiangiogenic domain of HRGP, exerts its effects through signal transduction targeting focal adhesions, thereby interrupting VEGF-induced endothelial cell motility.