Palladium(II) chloride
Synonym(s):
Palladium(II) chloride (59% Pd)
Empirical Formula (Hill Notation):
Cl2Pd
CAS Number:
Molecular Weight:
177.33
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Reaction Type
Reagent Type
Core
Special Grade
Showing 1-7 of 7
Compare
Product Number
Description
Pricing