Skip to Content
Merck
Carbon nanotube, multi-walled >98% carbon basis, O.D. × L 6-13 nm × 2.5-20 μm

Carbon nanotube, multi-walled

Carbon nanotube, multi-walled >98% carbon basis, O.D. × L 6-13 nm × 2.5-20 μm
Synonym(s):
MWCNT, MWNT, Multiwall carbon nanotube, DWNT, Double wall carbon nanotubes
CAS Number:

Products

Brand
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-13 of 13
Compare
Product Number
Description
Pricing