Skip to Content
Merck
All Photos(1)

Key Documents

4S8501

Supelco

Acrolein

analytical standard

Synonym(s):

2-Propenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCHO
CAS Number:
Molecular Weight:
56.06
Beilstein:
741856
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Agency

EPA 603,8240,8260
MISA (Canada) Group 18

vapor density

1.94 (vs air)

vapor pressure

4.05 psi ( 20 °C)

description

Separate Source

CofA

current certificate can be downloaded

autoignition temp.

428 °F

feature

standard type calibration

expl. lim.

31 %

packaging

pkg of 1 × 100 mg (4S8501)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.403 (lit.)

bp

53 °C (lit.)

mp

−87 °C (lit.)

solubility

H2O: soluble

density

0.839 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)C=C

InChI

1S/C3H4O/c1-2-3-4/h2-3H,1H2

InChI key

HGINCPLSRVDWNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acrolein is a carbonyl compound and a hazardous chemical, released into the environment through various manufacturing processes.

Application

Acrolein is used as an analytical reference standard for the quantification of the analyte in environmental matrices and biological samples using different chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of acrolein in human urine by headspace gas chromatography and mass spectrometry
Sakura N, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 719(1-2), 209-212 (1998)
Simple Determination of Acrolein in Surface and Drinking Water by Headspace SPME GC?MS
Lim H-H and Shin S-H
Chromatographia, 75(15-16), 943-948 (2012)
Klaus Abraham et al.
Molecular nutrition & food research, 55(9), 1277-1290 (2011-09-08)
Acrolein is an α,β-unsaturated aldehyde formed by thermal treatment of animal and vegetable fats, carbohydrates and amino acids. In addition it is generated endogenously. As an electrophile, acrolein forms adducts with gluthathione and other cellular components and is therefore cytotoxic.
Kiflai Bein et al.
Molecular nutrition & food research, 55(9), 1342-1360 (2011-10-14)
Acrolein is a respiratory irritant that can be generated during cooking and is in environmental tobacco smoke. More plentiful in cigarette smoke than polycyclic aromatic hydrocarbons (PAH), acrolein can adduct tumor suppressor p53 (TP53) DNA and may contribute to TP53-mutations
Richard M LoPachin et al.
Chemical research in toxicology, 22(9), 1499-1508 (2009-07-21)
Acrolein and 4-hydroxy-2-nonenal (HNE) are byproducts of lipid peroxidation and are thought to play central roles in various traumatic injuries and disease states that involve cellular oxidative stress, for example, spinal cord trauma, diabetes, and Alzheimer's disease. In this review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service