Skip to Content
Merck
All Photos(1)

Key Documents

47654-U

Supelco

Chloroacetic acid solution

certified reference material, 1000 μg/mL in methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 552.2

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in methyl tert-butyl ether

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C2H3ClO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

FOCAUTSVDIKZOP-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-27.4 °F - closed cup

Flash Point(C)

-33.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Wang Yanli et al.
Carbohydrate research, 344(13), 1764-1769 (2009-08-12)
Carboxymethyl starch (CMS) was obtained as a product of the reaction of starch and monochloroacetic acid (MCA) in the presence of sodium hydroxide. The influence of the molar ratio of NaOH/AGU, the molar ratio of MCA/AGU, the reaction time, reaction
[Investigation on coal work's production use of chloroacetic acid in one province].
Ying Bai et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 28(10), 781-782 (2010-12-04)
Rosanne Slingsby et al.
Journal of chromatographic science, 47(7), 523-528 (2009-09-24)
This paper presents the determination of nine haloacetic acids (HAAs) in high ionic strength, treated effluent waters using an ion chromatography-electrospray ionization-tandem mass spectrometry (IC-ESI-MS-MS) method with internal standards and discussions of each of the method parameters. Data is also
Xianbin Su et al.
BMC microbiology, 12, 267-267 (2012-11-22)
Acetate is a commonly used substrate for biosynthesis while monochloroacetate is a structurally similar compound but toxic and inhibits cell metabolism by blocking the citric acid cycle. In Burkholderia species MBA4 haloacetate was utilized as a carbon and energy source
Patrick Levallois et al.
Epidemiology (Cambridge, Mass.), 23(2), 267-276 (2012-02-10)
There is concern about possible effects of disinfection by-products on reproductive outcomes. The purpose of this study was to evaluate the association between maternal exposure to chlorination by-products and the risk of delivering a small for-gestational-age (SGA) neonate. We conducted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service