Skip to Content
Merck
All Photos(1)

Key Documents

47516-U

Supelco

Gasoline, premium unleaded

certified reference material, refinery grade, 20,000 μg/mL in methanol

Synonym(s):

Petrol, Gasoline, Petroleum benzin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

20,000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
petroleum

format

single component solution

storage temp.

2-30°C

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - Muta. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Dauger et al.
Archives de pediatrie : organe officiel de la Societe francaise de pediatrie, 10(3), 221-223 (2003-06-28)
The Authors report a case of acute White-Spirit poisoning with pulmonary hypertension associated to respiratory distress syndrome. A 14-month-old infant drank an unknown quantity of White-Spirit while his parents were painting. After he spontaneously vomited, he presented a seizure at
Lena Ernstgård et al.
Journal of applied toxicology : JAT, 29(3), 255-262 (2008-12-17)
A move from 'standard' white spirit (stdWS, 15-20% aromatics) to low-aromatic or dearomatized white spirit (deWS) has been seen, as the latter are considered to carry a smaller risk of health effects. However, data on health risks of deWS on
Gunnar Damgård Nielsen et al.
Basic & clinical pharmacology & toxicology, 98(2), 115-123 (2006-02-01)
Numerous studies have suggested that long-term occupational exposure to white spirit may cause chronic toxic encephalopathy (WHO 1996). This review summarizes the chronic nervous system effects of white spirit in animal studies during a 30-year period. First, routine histopathology was
P S Tofts et al.
Magnetic resonance imaging, 15(1), 69-75 (1997-01-01)
The selection of a suitable low dielectric constant liquid for radiofrequency coil nonuniformity measurements is described. Measurements of dielectric constant (relative permittivity) were made on a range of candidate liquids. After excluding liquids that were too inflammable or too viscous
Wan N S Mat-Desa et al.
Analytical chemistry, 83(20), 7745-7754 (2011-09-17)
Three different medium petroleum distillate (MPD) products (white spirit, paint brush cleaner, and lamp oil) were purchased from commercial stores in Glasgow, Scotland. Samples of 10, 25, 50, 75, 90, and 95% evaporated product were prepared, resulting in 56 samples

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service