Skip to Content
Merck
All Photos(1)

Key Documents

SRP5221

Sigma-Aldrich

KDM5C (1-671), GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DXS1272E, JARID1C, MRXJ, MRXSJ, XE169

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~118 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KDM5C(8242)

General description

KDM5C or lysine (K)-specific demethylase 5C is a member of the SMCY homolog family that encodes a protein with one ARID domain, one JmjC domain, one JmjN domain and two PHD-type zinc fingers. KDM5C is mainly expressed in brain and skeletal muscle. KDM5C is involved in the regulation of transcription and chromatin remodelling that cause X-linked mental retardation. KDM5C is also essential for spermatogenesis and expression of male-specific minor histocompatibility antigens.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A I Agulnik et al.
Human molecular genetics, 3(6), 873-878 (1994-06-01)
A new mouse Y chromosome gene, Smcy, has been isolated from the region encoding Spy, a spermatogenesis gene and Hya and Sdma, the genes that, respectively, control the expression of the male specific minor histocompatibility antigen H-Y, as measured by
Lars Riff Jensen et al.
American journal of human genetics, 76(2), 227-236 (2004-12-09)
In families with nonsyndromic X-linked mental retardation (NS-XLMR), >30% of mutations seem to cluster on proximal Xp and in the pericentric region. In a systematic screen of brain-expressed genes from this region in 210 families with XLMR, we identified seven

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service