Skip to Content
Merck
All Photos(1)

Key Documents

SRP4326

Sigma-Aldrich

Apo-D human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

ApoD, Apolipoprotein D

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

~20 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ApoD(347)

General description

Apolipoprotein D (APOD) is encoded by the gene mapped to human chromosome 3q29. The encoded protein is ubiquitously expressed in the body, but at high levels in glial cells of the brain, fibroblasts, spleen, testes and mammary cysts. APOD is a lipocalin containing an eight-stranded antiparallel β-sheet barrel-like structure forming a hydrophobic pocket.
Apolipoprotein-D (Apo-D) is mainly associated with high density lipoproteins in human plasma. It is expressed in numerous tissues having high levels of expression in spleen, testes and brain. Apo-D is a multi-ligand, multi-functional transporter and transports a ligand from 1 cell to another within an organ, scavenge a ligand within an organ for transport to the blood or could transport a ligand from the circulation to specific cells within a tissue. The recombinant human Apo-D expressed from E. coli is a single, non-glycosylated, Polypeptide chain containing 174 amino acids and having a molecular mass of 19.8 kDa.

Biochem/physiol Actions

Apolipoprotein D (APOD) plays an essential role in the development and repair of the brain and protection from oxidative stress. In addition, it also plays an essential role in lipid metabolism by facilitating the binding of HDL (high density lipoprotein) to LDL (low density lipoprotein) and to grow T24 carcinomas. Variation in the APOD expression acts as a genetic marker for obesity and several components of syndrome X.

Physical form

Lyophilized from 1 mg/ml in 4 mM KH2PO4, 16 mM Na2HPO4 and 115 mM NaCl pH7.5.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in dH2O to a working volume of 0.5 mg/mL and let the lyophilized pellet dissolve completely. This solution can then be diluted into other aqueous buffers and stored at 4°C for 1 week or –20°C for future use.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative-Trait Loci Influencing Body-Mass Index Reside on Chromosomes 7 and 13: The National Heart, Lung, and Blood Institute Family Heart Study
Feitosa M F, et al.
American Journal of Human Genetics, 70(1), 72-82 (2002)
ApoD Mediates Binding of HDL to LDL and to Growing T24 Carcinoma
Braesch-Andersen S, et al.
PLoS ONE, 9(12), e115180-e115180 (2014)
Apolipoprotein-D polymorphism: a genetic marker for obesity and hyperinsulinemia.
Vijayaraghavan S, et al.
The Journal of Clinical Endocrinology and Metabolism, 79(2), 568-570 (1994)
Emilie Picard et al.
Molecular vision, 14, 928-941 (2008-05-30)
Transferrin (Tf) expression is enhanced by aging and inflammation in humans. We investigated the role of transferrin in glial protection. We generated transgenic mice (Tg) carrying the complete human transferrin gene on a C57Bl/6J genetic background. We studied human (hTf)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service