Skip to Content
Merck
All Photos(1)

Key Documents

SML1012

Sigma-Aldrich

MN-64

≥98% (HPLC)

Synonym(s):

2-[4-(1-Methylethyl)phenyl]-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16O2
CAS Number:
Molecular Weight:
264.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C2=CC=CC=C2OC(C3=CC=C(C(C)C)C=C3)=C1

InChI

1S/C18H16O2/c1-12(2)13-7-9-14(10-8-13)18-11-16(19)15-5-3-4-6-17(15)20-18/h3-12H,1-2H3

InChI key

PYTOHIUBXSJKQH-UHFFFAOYSA-N

Biochem/physiol Actions

MN-64 is a very potent inhibitor of Tankyrase 1 and 2 (IC50 = 6 and 72 nM, respectively). The compound MN-64 blocks Wnt signaling in HEK293 cells containing a Wnt pathway reporter system.
MN-64 is a very potent inhibitor of Tankyrase 1 and 2.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sukyeong Kim et al.
Nature communications, 10(1), 4898-4898 (2019-10-28)
Osteoarthritis (OA) is a prevalent degenerative disease, which involves progressive and irreversible destruction of cartilage matrix. Despite efforts to reconstruct cartilage matrix in osteoarthritic joints, it has been a difficult task as adult cartilage exhibits marginal repair capacity. Here we

Related Content

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service