Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200113

Sigma-Aldrich

Anti-EDC4 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Ge-1, Anti-HEDLS (Human enhancer of decapping, large subunit), Anti-RCD-8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IP
WB
clone:
polyclonal
species reactivity:
human
citations:
3
technique(s):
immunoprecipitation (IP): 5-10 μg using lysates of HEK-293 cells over expressing human EDC4
western blot: 0.5-1 μg/mL using lysates of HEK-293 cells over expressing human EDC4

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~150 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysates of HEK-293 cells over expressing human EDC4
western blot: 0.5-1 μg/mL using lysates of HEK-293 cells over expressing human EDC4

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EDC4(23644)
mouse ... Edc4(234699)
rat ... Edc4(361399)

General description

Enhancer of mRNA decapping 4 (EDC4) contains an N-terminal WD40 repeat and a C-terminal domain, which interacts with decapping enzyme (Dcp2) and mediates EDC4 oligomerization and P-body localization.
The EDC4 gene is located on the human chromosome at 16q22.1.

Specificity

Anti-EDC4 (C-terminal) specifically recognizes human EDC4.

Application

Anti-EDC4 (C-terminal) antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Enhancer of mRNA decapping 4 (EDC4) associates with mRNA-decapping enzyme 2 (hDcp2) and stimulates hDcp2 activity in vitro as well as mediates the interaction between decapping enzymes (DCP1 and DCP2).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in “frost-free” freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The C-terminal region of Ge-1 presents conserved structural features required for P-body localization
Jinek M, et al.
RNA, 14(10), 1991-1998 (2008)
Erin J Gardiner et al.
Journal of psychiatric research, 47(4), 425-437 (2012-12-12)
Peripheral blood mononuclear cells (PBMCs) represent an accessible tissue source for gene expression profiling in schizophrenia that could provide insight into the molecular basis of the disorder. This study used the Illumina HT_12 microarray platform and quantitative real time PCR
Martin Fenger-Grøn et al.
Molecular cell, 20(6), 905-915 (2005-12-21)
Decapping is a key step in mRNA turnover. However, the composition and regulation of the human decapping complex is poorly understood. Here, we identify three proteins that exist in complex with the decapping enzyme subunits hDcp2 and hDcp1: hEdc3, Rck/p54

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service