Skip to Content
Merck
All Photos(1)

Key Documents

SAB2103187

Sigma-Aldrich

Anti-AHNAK2 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

85 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AHNAK2(113146)

Immunogen

Synthetic peptide directed towards the middle region of human AHNAK2

Application

Anti-AHNAK2 antibody produced in rabbit is suitable for western blot analysis.

Sequence

Synthetic peptide located within the following region: KEEGELIGPVGTGLDSRVMVTSAARTELILPEQDRKADDESKGSGLGPNEG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Sekiya et al.
The Journal of biological chemistry, 274(20), 13900-13907 (1999-05-13)
We have recently shown that phospholipase C-gamma (PLC-gamma) is activated by tau, a neuronal cell-specific microtubule-associated protein, in the presence of arachidonic acid. We now report that non-neuronal tissues also contain a protein that can activate PLC-gamma in the presence
Hannelore Haase
Cardiovascular research, 73(1), 19-25 (2006-10-19)
Ahnak, originally identified as a giant, tumour-related phosphoprotein, has emerged as an important signalling molecule in a wide range of physiological activities. In this article, current knowledge will be reviewed that places ahnak into the context of cardiac L-type Ca2+
Akihiko Komuro et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(12), 4053-4058 (2004-03-10)
To explore the function of the giant AHNAK molecule, first described in 1992 [Shtivelman, E., Cohen, F. E. & Bishop, J. M. (1992) Proc. Natl. Acad. Sci. USA 89, 5472-5476], we created AHNAK null mice by homologous recombination. Homozygous knockouts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service