Skip to Content
Merck
All Photos(1)

Key Documents

CPS2120

Sigma-Aldrich

Chemiluminescent Peroxidase Substrate for ELISA

sufficient for 120 mL substrate

Synonym(s):

CPS2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

form

liquid

usage

sufficient for 120 mL substrate

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Two component liquid substrate system with superior sensitivity, signal-to-noise, and linearity.

Kit Components Also Available Separately

Product No.
Description
SDS

  • Chemiluminescent Reagent 40 mL

  • Chemiluminescent Reaction Buffer 80 mL

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service