Skip to Content
Merck
All Photos(1)

Key Documents

B1883

Sigma-Aldrich

Benzidine

ISOPAC®, ≥98.0% (N)

Synonym(s):

4,4′-Diaminobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2
CAS Number:
Molecular Weight:
184.24
Beilstein:
742770
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.0% (N)

form

powder

SMILES string

Nc1ccc(cc1)-c2ccc(N)cc2

InChI

1S/C12H12N2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H,13-14H2

InChI key

HFACYLZERDEVSX-UHFFFAOYSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

Application

Benzidine (4,4′-Diaminobiphenyl) is an environmental genotoxin that posses increased risks of cancer to exposed people. Benzidine may be used as a reference material in procedures that analyze for benzidine. Benzidine can be used in studies on its mechanisms of genotoxicity.

Biochem/physiol Actions

Tumor initiator in mammalian liver and urinary bladder.

Packaging

Packaged in a 100 mL serum bottle with butyl rubber stopper and aluminum tear seal.

Caution

OSHA′s Safety and Health Standard 29 CFR 1910 does not apply to a solution or mixture containing less than 0.1%. Injecting a compatible solvent permits preparation of any desired strength solution without exposure to benzidine.

Reconstitution

Dissolving the contents in 100 mL of solvent yields a 0.1% solution.

Legal Information

Isopac is a registered trademark of Merck KGaA, Darmstadt, Germany

ISOPAC of

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Ling et al.
Biochemical and biophysical research communications, 401(4), 521-526 (2010-09-30)
Despite the significant role of S-ribosylhomocysteinase (LuxS) in the activated methyl cycle pathway and quorum sensing, the connectivity between luxS and other cellular functions remains incomplete. Herein, we show that luxS deletion significantly increases swimming motility and flagella synthesis in
Brian J Pak et al.
PLoS neglected tropical diseases, 8(8), e3002-e3002 (2014-08-08)
Strongyloidiasis is a persistent human parasitic infection caused by the intestinal nematode, Strongyloides stercoralis. The parasite has a world-wide distribution, particularly in tropical and subtropical regions with poor sanitary conditions. Since individuals with strongyloidiasis are typically asymptomatic, the infection can
G Choudhary
Chemosphere, 32(2), 267-291 (1996-01-01)
Benzidine, an odorless, white to slightly reddish-white crystalline organic compound, is an environmental contaminant that has been identified at about 30 National Priorities List (NPL) hazardous waste sites in the United States. In the environment, it is usually found attached
P D Josephy
Federation proceedings, 45(10), 2465-2470 (1986-09-01)
Benzidine oxidative activation may proceed by peroxidase-catalyzed one-electron oxidation via free radical intermediates, or by N-acetylation followed by monooxygenase-catalyzed N-hydroxylation. The peroxidase route has been examined by using horseradish peroxidase or prostaglandin H synthase in vitro. In the presence of
J Whysner et al.
Pharmacology & therapeutics, 71(1-2), 107-126 (1996-01-01)
The aromatic amine benzidine (BZ) has produced various tumors, including liver tumors, in mice, rats and hamsters. BZ forms DNA adducts in rodent liver, and it is positive in most genotoxicity tests. Only bladder tumors are produced in dogs and

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service