Skip to Content
Merck
All Photos(1)

Key Documents

AV46665

Sigma-Aldrich

Anti-ZMPSTE24 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FACE-1, Anti-FACE1, Anti-FLJ14968, Anti-STE24, Anti-Ste24p, Anti-Zinc metallopeptidase (STE24 homolog, S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

horse, rabbit, human, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the N terminal region of human ZMPSTE24

Sequence

Synthetic peptide located within the following region: LYSETEGTLILLFGGIPYLWRLSGRFCGYAGFGPEYEITQSLVFLLLATL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahid Ahmad et al.
American journal of medical genetics. Part A, 152A(11), 2703-2710 (2010-09-04)
Mandibuloacral dysplasia (MAD) is an autosomal recessive disorder characterized by hypoplasia of the mandible and clavicles, acro-osteolysis, and lipodystrophy due to mutations in LMNA or ZMPSTE24. Only six MAD patients are reported so far with ZMPSTE24 mutations and limited phenotypic
Alberto M Pendás et al.
Nature genetics, 31(1), 94-99 (2002-03-30)
The mouse ortholog of human FACE-1, Zmpste24, is a multispanning membrane protein widely distributed in mammalian tissues and structurally related to Afc1p/ste24p, a yeast metalloproteinase involved in the maturation of fungal pheromones. Disruption of the gene Zmpste24 caused severe growth
J M Freije et al.
Genomics, 58(3), 270-280 (1999-06-22)
Two human cDNAs encoding proteins similar to yeast enzymes involved in proteolytic processing of farnesylated proteins like a-factor mating pheromone and Ras2p have been cloned from an ovary cDNA library. These proteins have been tentatively called Face-1 and Face-2 (farnesylated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service