Skip to Content
Merck
All Photos(1)

Key Documents

92609

Sigma-Aldrich

N,N-Dimethylsphingosine

≥98.0% (TLC)

Synonym(s):

(2S,3R,4E)-2-(Dimethylamino)-4-octadecene-1,3-diol, N,N-Dimethyl-D-erythro-sphingosine, N,N-Dimethylsphingenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H41NO2
CAS Number:
Molecular Weight:
327.55
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥98.0% (TLC)

form

paste

composition

carbon content, 71.9-74.8%
nitrogen content, 4.1-4.5%

lipid type

sphingolipids

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\[C@H](O)[C@@H](CO)N(C)C

InChI

1S/C20H41NO2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)19(18-22)21(2)3/h16-17,19-20,22-23H,4-15,18H2,1-3H3/b17-16+/t19-,20+/m1/s1

InChI key

YRXOQXUDKDCXME-QWKHPXNBSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kimura et al.
Biochemical pharmacology, 44(8), 1585-1595 (1992-10-20)
Neutrophils display three major functions: (i) oxidative burst, (ii) phagokinetic activity, and (iii) trans-endothelial migration. Sphingosine (SPN) is known to inhibit oxidative burst in human neutrophils via inhibition of protein kinase C (PKC). SPN is metabolically converted into N,N-dimethylsphingosine (DMS)
K Endo et al.
Cancer research, 51(6), 1613-1618 (1991-03-15)
Sphingosine (SPN) has been claimed to be a negative modulator of transmembrane signaling through protein kinase C (PK-C) or some yet unidentified mechanism [for review see Y. A. Hannun and R. M. Bell, Science (Washington DC), 243: 500-507, 1989]. N,N-Dimethylsphingosine
Y Igarashi et al.
The Journal of biological chemistry, 265(10), 5385-5389 (1990-04-05)
Our previous study suggested that N,N-dimethylsphingosine, but not unsubstituted sphingosine, could be a modulator of protein kinase C in epidermoid carcinoma A431 cells, since N,N-dimethyl-D-erythrosphingenine showed a stronger stereospecific effect on protein kinase C activity in comparison with N,N-dimethyl-L-erythrosphingenine, unsubstituted
Lei Xiu et al.
The American journal of pathology, 185(2), 387-398 (2014-11-29)
Sphingosine 1-phosphate (S1P) is involved in multiple pathological processes, including fibrogenesis. S1P participates in mouse liver fibrogenesis via a paracrine manner. Herein, we investigated the involvement of S1P in human liver fibrosis. Human fibrotic samples were obtained from livers of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service