Skip to Content
Merck
All Photos(1)

Key Documents

P3600000

Propranolol hydrochloride for performance test

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Propranolol hydrochloride, (±)-1-Isopropylamino-3-(1-naphthyloxy)-2-propanol hydrochloride, DL-Propranolol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO2 · HCl
CAS Number:
Molecular Weight:
295.80
Beilstein:
4164259
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CC(C)NCC(O)COc1cccc2ccccc12

InChI

1S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18H,10-11H2,1-2H3;1H

InChI key

ZMRUPTIKESYGQW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Propranolol hydrochloride for performance test EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy C Arnold et al.
Neurology, 80(21), 1927-1933 (2013-04-26)
To determine the effect of low-dose propranolol on maximal exercise capacity in patients with postural tachycardia syndrome (POTS). We compared the effect of placebo vs a single low dose of propranolol (20 mg) on peak oxygen consumption (VO2max), an established
Philip M Short et al.
American journal of respiratory and critical care medicine, 187(12), 1308-1314 (2013-04-19)
Unblinded studies have shown improvements in airway hyperresponsiveness with chronic nadolol in steroid-naive patients with asthma. To assess the effects of chronic nonselective β-blockade as add-on to inhaled corticosteroids (ICS) in patients with asthma. A double-blind randomized placebo-controlled crossover trial
Nicole D Powell et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(41), 16574-16579 (2013-09-26)
Across a variety of adverse life circumstances, such as social isolation and low socioeconomic status, mammalian immune cells have been found to show a conserved transcriptional response to adversity (CTRA) involving increased expression of proinflammatory genes. The present study examines
Massimo Dal Monte et al.
Experimental eye research, 111, 27-35 (2013-03-27)
The mouse model of oxygen-induced retinopathy (OIR) is a well-established model of retinopathy of prematurity (ROP), characterized by the abnormal formation of new blood vessels, which is similar to ROP. In this model, we have recently shown that subcutaneous (sc)
Silvia Franzellitti et al.
Aquatic toxicology (Amsterdam, Netherlands), 140-141, 249-256 (2013-07-09)
Aquatic organisms are unintentionally exposed to a large number of pharmaceutical residues in their natural habitats. Ecotoxicological studies have agreed that these compounds are not harmful to aquatic organisms, as their environmental concentrations are typically too low. However, recent reports

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service