93066
Cinnamyl alcohol
analytical standard
Synonym(s):
3-Phenyl-2-propen-1-ol
About This Item
Recommended Products
grade
analytical standard
Quality Level
vapor density
4.6 (vs air)
vapor pressure
<0.01 mmHg ( 25 °C)
Assay
≥96.0% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
bp
250 °C (lit.)
mp
30-33 °C (lit.)
density
1.044 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
format
neat
storage temp.
2-8°C
SMILES string
OC\C=C\c1ccccc1
InChI
1S/C9H10O/c10-8-4-7-9-5-2-1-3-6-9/h1-7,10H,8H2/b7-4+
InChI key
OOCCDEMITAIZTP-QPJJXVBHSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
258.8 °F - closed cup
Flash Point(C)
126 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service