Skip to Content
Merck
All Photos(1)

Key Documents

80827

Supelco

N,N-Dimethyl-2-sulfamoylnicotinamide

PESTANAL®, analytical standard

Synonym(s):

2-(Aminosulfonyl)-N,N-dimethyl-3-pyridinecarboxamide, 2-Aminosulfonyl-N,N-dimethylnicotinamide, N,N-Dimethyl-2-sulfamoylpyridine-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N3O3S
CAS Number:
Molecular Weight:
229.26
Beilstein:
9193761
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

O=C(N(C)C)C1=CC=CN=C1S(=O)(N)=O

InChI

1S/C8H11N3O3S/c1-11(2)8(12)6-4-3-5-10-7(6)15(9,13)14/h3-5H,1-2H3,(H2,9,13,14)

InChI key

WYFKZPLSYVJLRB-UHFFFAOYSA-N

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsotne Chitiashvili et al.
Nature cell biology, 22(12), 1436-1446 (2020-12-02)
X-chromosome dosage compensation in female placental mammals is achieved by X-chromosome inactivation (XCI). Human pre-implantation embryos are an exception, in which dosage compensation occurs by X-chromosome dampening (XCD). Here, we examined whether XCD extends to human prenatal germ cells given
María Expósito-Serrano et al.
Current biology : CB, 30(16), 3212-3222 (2020-06-06)
An important question in cell biology is how cellular organelles partition during cell division. In organisms undergoing closed mitosis, the elongation of an intranuclear spindle drives nuclear division, generating two identically sized nuclei [1, 2]. However, how the site of
Diogo R Feleciano et al.
Frontiers in aging neuroscience, 11, 9-9 (2019-02-15)
A functional protein quality control machinery is crucial to maintain cellular and organismal physiology. Perturbation in the protein homeostasis network can lead to the formation of misfolded and aggregated proteins that are a hallmark of protein conformational disorders and aging.
Mette C Jørgensen et al.
Development (Cambridge, England), 145(17) (2018-08-11)
Mutations in Hes1, a target gene of the Notch signalling pathway, lead to ectopic pancreas by a poorly described mechanism. Here, we use genetic inactivation of Hes1 combined with lineage tracing and live imaging to reveal an endodermal requirement for
Philipp Trepte et al.
Molecular systems biology, 14(7), e8071-e8071 (2018-07-13)
Information on protein-protein interactions (PPIs) is of critical importance for studying complex biological systems and developing therapeutic strategies. Here, we present a double-readout bioluminescence-based two-hybrid technology, termed LuTHy, which provides two quantitative scores in one experimental procedure when testing binary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service