Skip to Content
Merck
All Photos(2)

Key Documents

79184

Supelco

Phenylbutazone

analytical standard

Synonym(s):

4-Butyl-1,2-diphenyl-3,5-pyrazolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N2O2
CAS Number:
Molecular Weight:
308.37
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

mp

106-108 °C (lit.)

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

15-25°C

SMILES string

CCCCC1C(=O)N(N(C1=O)c2ccccc2)c3ccccc3

InChI

1S/C19H20N2O2/c1-2-3-14-17-18(22)20(15-10-6-4-7-11-15)21(19(17)23)16-12-8-5-9-13-16/h4-13,17H,2-3,14H2,1H3

InChI key

VYMDGNCVAMGZFE-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Aarbakke
Clinical pharmacokinetics, 3(5), 369-380 (1978-09-01)
More than 25 years after phenylbutazone was introduced as a non-steroidal anti-inflammatory agent, basic knowledge is still accumulating on its pharmacokinetics in man. Phenylbutazone is almost completely absorbed after oral administration. A large fraction of the drug in plasma is
Robert D Combes
Alternatives to laboratory animals : ATLA, 41(3), 235-248 (2013-08-27)
Published in silico, in vitro, in vivo laboratory animal and human data, together with information on biotransformation and data from structure-activity analyses with two decision-tree systems (ACToR and Toxtree), have been used in a weight-of-evidence (WoE) assessment to determine whether
T Tobin et al.
Journal of veterinary pharmacology and therapeutics, 9(1), 1-25 (1986-03-01)
Phenylbutazone is an acidic, lipophilic, non-steroidal anti-inflammatory drug (NSAID). It is extensively metabolized in the horse. The metabolites so far identified, oxyphenbutazone, gamma-hydroxyoxyphenbutazone, account for some 25-30% of administered dose over 24 h. The plasma half-life of phenylbutazone and termination
[Multiple manifestations of intolerance in association with anti-inflammatory agents. Role of phenylbutazone].
F Tolot et al.
Semaine des hopitaux. Therapeutique, 52(3), 185-190 (1976-03-01)
M Maciazek-Jurczyk et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 74(1), 1-9 (2009-07-21)
The monitoring of drug concentration in blood serum is necessary in multi-drug therapy. Mechanism of drug binding with serum albumin (SA) is one of the most important factors which determine drug concentration and its transport to the destination tissues. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service