Skip to Content
Merck
All Photos(1)

Key Documents

77647

Supelco

Redox standard

400 mV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

description

redox potential: 400 mV ±5 mV, 25°C

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
pharmaceutical

format

mixture

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The value is the potential of a platinum electrode versus an Ag/AgCl reference in 3M potassium chloride

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service