Skip to Content
Merck
All Photos(1)

Key Documents

74139

Supelco

2-Ethylthiophene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8S
CAS Number:
Molecular Weight:
112.19
Beilstein:
105402
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.3% water

refractive index

n20/D 1.512-1.514
n20/D 1.513 (lit.)

bp

132-134 °C (lit.)

density

0.99 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCc1cccs1

InChI

1S/C6H8S/c1-2-6-4-3-5-7-6/h3-5H,2H2,1H3

InChI key

JCCCMAAJYSNBPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylthiophene is an organosulfur compound belonging to the class of alkylated thiophenes.

Application

2-Ethylthiophene may be used as an analytical reference standard for the determination of the analyte in wastewater samples, diesel particulate matter and diesel fuel and industrial cooked ham by various chromatography techniques.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

85.3 °F - closed cup - (own results)

Flash Point(C)

29.6 °C - closed cup - (own results)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of polycyclic aromatic sulfur heterocycles in diesel particulate matter and diesel fuel by gas chromatography with atomic emission detection
Liang F, et al.
Journal of Chromatography A, 1114(1), 145-153 (2006)
Identification and origin of odorous sulfur compounds in cooked ham
Thomas C, et al.
Food Chemistry, 155, 207-213 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service