Skip to Content
Merck
All Photos(1)

Key Documents

68223

Supelco

2,4-Lutidine

analytical standard

Synonym(s):

2,4-Dimethylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Beilstein:
1506
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.3% water

refractive index

n20/D 1.498-1.502
n20/D 1.499 (lit.)

bp

159 °C (lit.)

mp

−60 °C (lit.)

density

0.927 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cc1ccnc(C)c1

InChI

1S/C7H9N/c1-6-3-4-8-7(2)5-6/h3-5H,1-2H3

InChI key

JYYNAJVZFGKDEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service