Skip to Content
Merck
All Photos(1)

Key Documents

62728

Supelco

(2RS)-2-[4-(2-Methylpropyl)phenyl]propan-1-ol

pharmaceutical impurity standard

Synonym(s):

Ibuprofen impurity P (PhEur), β-Methyl-4-(2-methylpropyl)benzeneethanol, 2-(4-Isobutylphenyl)propanol, 2-(4-Isobutylphenyl)propyl alcohol, Ibuprofen alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
Beilstein:
1945151
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OCC(C)C1=CC=C(CC(C)C)C=C1

InChI

1S/C13H20O/c1-10(2)8-12-4-6-13(7-5-12)11(3)9-14/h4-7,10-11,14H,8-9H2,1-3H3

InChI key

IZXWIWYERZDWOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service