Skip to Content
Merck
All Photos(1)

Key Documents

61382

Supelco

D-α-Hydroxyglutaric acid disodium salt

analytical standard

Synonym(s):

D-2HG, (R)-2-Hydroxypentanedioic acid disodium salt, Disodium (R)-2-hydroxyglutarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H6O5Na2
CAS Number:
Molecular Weight:
192.08
Beilstein:
5318041
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥98.0% (GC)

optical activity

[α]/D 8.5±1.5°, c = 1 in 0.1 M NaOH

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

[Na].O[C@H](CCC(O)=O)C(O)=O

InChI

1S/C5H8O5.2Na/c6-3(5(9)10)1-2-4(7)8;;/h3,6H,1-2H2,(H,7,8)(H,9,10);;/q;2*+1/p-2/t3-;;/m1../s1

InChI key

DZHFTEDSQFPDPP-HWYNEVGZSA-L

Looking for similar products? Visit Product Comparison Guide

Application

D-α-Hydroxyglutaric acid disodium salt may be used as an analytical standard for the quantification of 2-hydroxyglutarate (2HG) and 2-hydroxyglutarate (2HG) enantiomers in human serum samples of patients with isocitrate dehydrogenase (IDH) mutated tumors using chromatography-based techniques.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of 2-hydroxyglutarate enantiomers in serum by chiral gas chromatography-tandem mass spectrometry and its application as a biomarker for IDH mutant gliomas
Strain KS, et al.
Clinical mass spectrometry, 15, 16-25 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service