Skip to Content
Merck
All Photos(2)

Key Documents

51704

Supelco

Tanshinone IIA

analytical standard

Synonym(s):

1,6,6-Trimethyl-6,7,8,9-tetrahydrophenanthro[1,2-b]furan-10,11-dione, Dan Shen ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18O3
CAS Number:
Molecular Weight:
294.34
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

204-208 °C

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Cc1coc2-c3ccc4c(CCCC4(C)C)c3C(=O)C(=O)c12

InChI

1S/C19H18O3/c1-10-9-22-18-12-6-7-13-11(5-4-8-19(13,2)3)15(12)17(21)16(20)14(10)18/h6-7,9H,4-5,8H2,1-3H3

InChI key

HYXITZLLTYIPOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Phenanthrenequinone constituent of Chinese medicinal herb Danshen (Salvia miltiorrhiza). Anti-inflammatory. Antioxidant. Cytotoxic against a variety of cell lines, inlcuding human glioma cells.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Si Gao et al.
Atherosclerosis, 220(1), 3-10 (2011-07-22)
Tanshinone IIA (TS), a pharmacologically active component isolated from the rhizome of the Chinese herb Salvia miltiorrhiza Bunge (Danshen), has been clinically used in Asian countries for the prevention and treatment of coronary heart disease. Recently, the pharmacological properties of
Hui Ma et al.
The American journal of Chinese medicine, 41(1), 197-210 (2013-01-23)
Natural product Tanshinone IIA (TanIIA) induces apoptosis and differentiation in hepatocellular carcinoma (HCC) cells, but its clinical use is limited due to poor water solubility and lack of appropriate formulations for drug delivery. In this study, we capsulated TanIIA into
Xiao Zhang et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 48(4-5), 595-603 (2013-01-29)
The aim of this study was to develop novel multiple agents loaded poly (D,L-lactide-co-glycolide acid) (PLGA) nanoparticles (NPs) and evaluate their potential for brain delivery via inner ear administration. PLGA NPs loaded with salvianolic acid B (Sal B), tanshinone IIA
Paul Chan et al.
The American journal of Chinese medicine, 40(6), 1307-1319 (2012-12-12)
The injury of endothelial cell is the critical event of vascular disease. In endothelial cell, oxidative stress is regarded as critical to pathogenic factors in endothelial cell injury and apoptosis. Tanshinone IIA is the main effective component of Salvia miltiorrhiza
Xiao-Hong Tian et al.
Expert opinion on therapeutic patents, 23(1), 19-29 (2012-10-26)
Tanshinone IIA (TSIIA), cryptotanshinone and tanshinone I are major bioactive constituents of Danshen, a Chinese herbal medicine. Sodium tanshinone IIA sulfonate (STS) is a water-soluble derivative of TSIIA and it is currently used for treating cardiovascular diseases in China. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service