51512
Ammonium sulfamate
PESTANAL®, analytical standard
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
Assay
99.0-101.0% (RT)
shelf life
limited shelf life, expiry date on the label
loss
≤1% loss on drying, 110°C
mp
131-135 °C (lit.)
solubility
H2O: 0.5 g/10 mL, clear, colorless
application(s)
agriculture
environmental
format
neat
SMILES string
N.NS(O)(=O)=O
InChI
1S/H3NO3S.H3N/c1-5(2,3)4;/h(H3,1,2,3,4);1H3
InChI key
GEHMBYLTCISYNY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service