About This Item
Recommended Products
grade
analytical standard
Quality Level
vapor density
4.9 (vs air)
vapor pressure
2.9 mmHg ( 20 °C)
Assay
≥98.0% (GC)
autoignition temp.
356 °F
shelf life
limited shelf life, expiry date on the label
expl. lim.
5.6 %
refractive index
n20/D 1.415-1.419
n20/D 1.416 (lit.)
bp
155-158 °C (lit.)
40-42 °C/11 mmHg (lit.)
mp
−93.5 °C (lit.)
density
0.753 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
food and beverages
personal care
SMILES string
CCCN(CCC)CCC
InChI
1S/C9H21N/c1-4-7-10(8-5-2)9-6-3/h4-9H2,1-3H3
InChI key
YFTHZRPMJXBUME-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
93.2 °F - closed cup
Flash Point(C)
34 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service