Skip to Content
Merck
All Photos(1)

Key Documents

49679

Supelco

p-Cymene

certified reference material, TraceCERT®

Synonym(s):

1-Isopropyl-4-methylbenzene, 4-Isopropyltoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH(CH3)2
CAS Number:
Molecular Weight:
134.22
Beilstein:
1903377
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

4.62 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)
3.7 mmHg ( 37.7 °C)

autoignition temp.

817 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

5.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.490 (lit.)

bp

176-178 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(C)c1ccc(C)cc1

InChI

1S/C10H14/c1-8(2)10-6-4-9(3)5-7-10/h4-8H,1-3H3

InChI key

HFPZCAJZSCWRBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Lydia E H Paul et al.
Inorganic chemistry, 51(2), 1057-1067 (2012-01-10)
The relative affinity of the cationic triangular metallaprism, [(pCH(3)C(6)H(4)Pr(i))(6)Ru(6)(tpt)(2)(dhbq)(3)](6+) ([1](6+)), for various amino acids, ascorbic acid, and glutathione (GSH) has been studied at 37 °C in aqueous solutions at pD 7, using NMR spectroscopy and electrospray ionization mass spectrometry (ESI-MS).
Vanessa Kar-Yan Lo et al.
Journal of the American Chemical Society, 134(18), 7588-7591 (2012-04-26)
Complex [(p-cymene)Ru(η(1)-O(2)CCF(3))(2)(OH(2))] mediated transformation of α-diazoacetamides ArCH(2)N(C(CH(3))(3))C(O)CHN(2) to result in carbene insertion into the primary C-H bond exclusively, with the γ-lactam products being isolated in up to 98% yield. This unexpected reaction is striking in view of the presence of
Natalia Busto et al.
Chemistry, an Asian journal, 7(4), 788-801 (2012-02-07)
The reactions of two diaminotriazine ligands 2,4-diamino-6-(2-pyridyl)-1,3,5-triazine (2-pydaT) and 6-phenyl-2,4-diamino-1,3,5-triazine (PhdaT) with ruthenium-arene precursors led to a new family of ruthenium(II) compounds that were spectroscopically characterized. Four of the complexes were cationic, with the general formula [(η(6)-arene)Ru(κ(2)-N,N-2-pydaT)Cl]X (X=BF(4), TsO; arene=p-cymene:
Lutz Ackermann et al.
Organic letters, 14(8), 2146-2149 (2012-04-13)
The ruthenium(II) carboxylate complex [Ru(O(2)CMes)(2)(p-cymene)] enabled efficient direct arylations of unactivated C-H bonds with easily available, inexpensive phenols. Extraordinary chemoselectivity of the well-defined ruthenium catalyst set the stage for challenging C-H/C-O bond functionalizations to occur under solvent-free conditions as well
Kenichi Ogata et al.
Dalton transactions (Cambridge, England : 2003), 42(7), 2362-2365 (2012-12-14)
A series of 1,2,3-triazol-5-ylidene (tzNHC) complexes of a (p-cymene)ruthenium system was synthesized. The reactivity of the N-bonded and C-bonded aryl groups in the tzNHC ligand was found to be significantly different with respect to intramolecular aromatic C-H bond activation.

Protocols

Cinnamoµm Camphor, also known as the camphor tree, is grown in several parts of the world including Japan, Taiwan, and Australia. Camphor, which is used in medicinal and cosmetic preparations, is derived from the leaves and wood of this tree.

-Cymene; (−)-Menthone; α-Terpineol, natural, ≥96%, FCC, FG; Terpinolene; β-Bourbonene; 1-Octen-3-ol; β-Caryophyllene; Linalool; α-Terpinene; (−)-Menthol

-Cymene; Linalool; Menthol; α-Terpineol; Menthyl acetate

Cymene; 4,5,6,7-Tetrahydro-3,6-dimethylbenzofuran; Linalool; Menthol; Menthone; Menthyl acetate; Germacrene D; Bicyclogermacrene; Thymol

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service