Skip to Content
Merck
All Photos(1)

Key Documents

46297

Supelco

Furazolidone

VETRANAL®, analytical standard

Synonym(s):

3-(5-Nitrofurfurylideneamino)-2-oxazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3O5
CAS Number:
Molecular Weight:
225.16
Beilstein:
8317414
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-positive bacteria
parasites

application(s)

cleaning products
clinical
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

Mode of action

enzyme | inhibits

SMILES string

[O-][N+](=O)c1ccc(\C=N\N2CCOC2=O)o1

InChI

1S/C8H7N3O5/c12-8-10(3-4-15-8)9-5-6-1-2-7(16-6)11(13)14/h1-2,5H,3-4H2/b9-5+

InChI key

PLHJDBGFXBMTGZ-WEVVVXLNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Furazolidone is an effective antiprotozoal and antibacterial agent.

Application

Furazolidone may be used as a reference standard:
  • For the determination of the analyte in pharmaceutical formulations by second-derivative spectrophotometry.
  • For the determination of the analyte in animal feeds using liquid chromatography with ultraviolet and thermospray mass spectrometric detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Formulation and advantages of furazolidone in liposomal drug delivery systems.
Alam MI, et al.
European Journal of Pharmaceutical Sciences, 84, 139- 145 (2016)
Determination of furazolidone in animal feeds using liquid chromatography with UV and thermospray mass spectrometric detection.
McCracken RJ and Kennedy DG
Journal of Chromatography A, 771(1-2), 349-354 (1997)
C V Prasad et al.
Journal of pharmaceutical and biomedical analysis, 21(5), 961-968 (2000-03-07)
A second-derivative spectrophotometric procedure has been developed for the simultaneous determination of tinidazole (TD), furazolidone (FD) and diloxanide furoate (DF) in a commercial preparation. The method consists of the utilization of second-derivative absorption spectra of tablet extract in distilled water
André Gustavo Tempone et al.
International journal of antimicrobial agents, 36(2), 159-163 (2010-06-18)
Drug delivery systems are promising pharmaceutical formulations used to improve the therapeutic index of drugs. In this study, we developed a liposomal formulation of furazolidone that targets Leishmania (Leishmania) chagasi amastigotes in a hamster model. Using laser scanning confocal microscopy
Xi Jin et al.
Toxicology letters, 201(3), 205-212 (2011-01-05)
Furazolidone (FZD) is an antimicrobial agent that has been shown to have mutagenic, genotoxic and potentially carcinogenic properties when tested in a variety of systems in vitro and in vivo. In this study, we investigated FZD's DNA damaging effect in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service