Skip to Content
Merck
All Photos(2)

Key Documents

45432

Supelco

Dichlofenthion

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13Cl2O3PS
CAS Number:
Molecular Weight:
315.15
Beilstein:
1993440
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1ccc(Cl)cc1Cl

InChI

1S/C10H13Cl2O3PS/c1-3-13-16(17,14-4-2)15-10-6-5-8(11)7-9(10)12/h5-7H,3-4H2,1-2H3

InChI key

WGOWCPGHOCIHBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongbo Li et al.
Analytical chemistry, 83(13), 5290-5295 (2011-05-25)
A novel photoelectrochemical sensor for detection of the organophosphorus pesticide (OP) dichlofenthion using nanometer-sized titania coupled with a screen-printed electrode is presented. Nonelectroactive dichlofenthion can be indirectly determined through the photocatalytical degradation of dichlofenthion with nanometer-sized titania. The electrochemical characterization
M O'Flynn et al.
Australian veterinary journal, 56(2), 67-69 (1980-02-01)
A survey was conducted in 1977-78 to determine the level of insecticide resistance in field strains of Lucilia cuprina from a wide area of Queensland. Discriminating dose data showed that all 19 strains tested were resistant to diazinon, dichlofenthion, fenthion-ethyl
I D Perkins
Australian veterinary journal, 64(1), 17-20 (1987-01-01)
Some insecticides to control and prevent screw-worm fly strike by Chrysomya bezziana in calves and adult cattle were tested in field trials on cattle in Sabah, East Malaysia. Ivermectin injected subcutaneously in newborn calves at 200 mu/kg provided 10 days
Ivan Notardonato et al.
Analytical and bioanalytical chemistry, 410(14), 3285-3296 (2018-03-20)
Baby foods are either a soft, liquid paste or an easily chewed food since babies lack developed muscles and teeth to chew effectively. Babies typically move to consuming baby food once nursing or formula is not sufficient for the child's
Jishi Wang et al.
Journal of chromatography. A, 1521, 10-18 (2017-09-25)
In this research, the manual two-step QuEChERS approach has been streamlined and automated into a one-step method using a cleanup tube fitted within an extraction tube. A novel automatic QuEChERS combination have been developed to simplify the QuEChERS procedures and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service