42899
(+)-Isocorydine hydrochloride
analytical standard
Synonym(s):
11-Hydroxy-1,2,10-trimethoxyaporphine, Isocorydine hydrochloride, Luteanine
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥98.0% (HPLC)
optical activity
[α]/D 180±10°, c = 0.1 in methanol
shelf life
limited shelf life, expiry date on the label
impurities
≤5.0% water
mp
215-218 °C (dec.) (lit.)
application(s)
food and beverages
format
neat
SMILES string
OC1=C2C(C[C@@]3([H])C4=C2C(OC)=C(OC)C=C4CCN3C)=CC=C1OC.Cl
InChI
1S/C20H23NO4.ClH/c1-21-8-7-12-10-15(24-3)20(25-4)18-16(12)13(21)9-11-5-6-14(23-2)19(22)17(11)18;/h5-6,10,13,22H,7-9H2,1-4H3;1H/t13-;/m0./s1
InChI key
ZWSKLEMBDRWSNZ-ZOWNYOTGSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service