Skip to Content
Merck
All Photos(1)

Key Documents

41557

Supelco

Sesamol

analytical standard

Synonym(s):

3,4-(Methylenedioxy)phenol, 5-Benzodioxolol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O3
CAS Number:
Molecular Weight:
138.12
Beilstein:
127405
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

mp

62-65 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

Oc1ccc2OCOc2c1

InChI

1S/C7H6O3/c8-5-1-2-6-7(3-5)10-4-9-6/h1-3,8H,4H2

InChI key

LUSZGTFNYDARNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sesamol, a principal phytophenol of sesame oil lignans, is typically used as a dietary compound.(10)

Application

Sesamol may be used as an analytical standard for the determination of the analyte in foods, nutraceutical products, sesame cake extract as well as sesame oil and oil blends by various analytical techniques.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sesamol: a powerful functional food ingredient from sesame oil for cardioprotection
Jayaraj P, et al.
Food & Function, 11(2) (2020)
Encapsulation of sesamol in phosphatidyl choline micelles: Enhanced bioavailability and anti-inflammatory activity
Yashaswini PS, et al.
Food Chemistry, 228(7), 330-337 (2017)
Free radical scavenging behavior of antioxidant compounds of sesame (Sesamum indicum L.) in DPPH? system
Suja K, et al.
Journal of Agricultural and Food Chemistry, 52(4), 912-915 (2004)
Studies on quality of coconut oil blends after frying potato chips
Khan KI, et al.
Journal of the American Oil Chemists' Society, 85(12), 1165-1172 (2008)
Identification of sesamol byproducts produced by plasma treatment with inhibition of advanced glycation endproducts formation and ONOO-scavenging activities
Jeong YH, et al.
Food Chemistry, 53(7), 126196-126196 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service