Skip to Content
Merck
All Photos(1)

Key Documents

37012

Supelco

Beauvericin

VETRANAL®, analytical standard

Synonym(s):

cyclo(D-α-Hydroxyisovaleryl-L-N-methyl-Phe)3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H57N3O9
CAS Number:
Molecular Weight:
783.95
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CC(C)C1OC(=O)C(Cc2ccccc2)N(C)C(=O)C(OC(=O)C(Cc3ccccc3)N(C)C(=O)C(OC(=O)C(Cc4ccccc4)N(C)C1=O)C(C)C)C(C)C

InChI

1S/C45H57N3O9/c1-28(2)37-40(49)46(7)35(26-32-21-15-11-16-22-32)44(53)56-39(30(5)6)42(51)48(9)36(27-33-23-17-12-18-24-33)45(54)57-38(29(3)4)41(50)47(8)34(43(52)55-37)25-31-19-13-10-14-20-31/h10-24,28-30,34-39H,25-27H2,1-9H3

InChI key

GYSCAQFHASJXRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Beauvericin is a mycotoxin mainly produced by several fungi belonging to Fusarium genus, generally occurring in agricultural commodities.

Application

Beauvericin may be used as analytical reference standard for the quantification of the analyte in cultures of Fusarium subglutinans, naturally contaminated maize, and biological samples using various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Reconstitution

Dried down, ~100μg/mL after reconstitution with 1 mL of water

Analysis Note

purity : ≥92.5% (HPLC)

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of beauvericin, enniatins, and fusaproliferin by high performance liquid chromatography
Monti.MS, et al.
Journal of Agricultural and Food Chemistry, 48(8), 3317-3320 (2000)
Determination of the Fusarium mycotoxins, fusaproliferin and beauvericin by high-performance liquid chromatography-electrospray ionization mass spectrometry
Sewram V, et al.
Journal of Chromatography A, 858(2), 175-185 (1999)
Quantitative determination of the Fusarium mycotoxins beauvericin, enniatin A, A1, B and B1 in pig plasma using high performance liquid chromatography--tandem mass spectrometry
Devreese M, et al.
Talanta, 106(2), 212-219 (2013)
Anton A Tonshin et al.
Toxicology, 276(1), 49-57 (2010-07-14)
The mechanisms of cell toxicity of mycotoxins of the enniatin family produced by Fusarium sp. enniatin B, a mixture of enniatin homologues (3% A, 20% A(1), 19% B, 54% B1) and beauvericin, were investigated. In isolated rat liver mitochondria, exposure
A S Ficheux et al.
Toxicon : official journal of the International Society on Toxinology, 71, 1-10 (2013-05-21)
The aim of this study was to assess the in vitro effects of emerging mycotoxins beauvericin, enniatin B and moniliformin on human dendritic cells and macrophages. Beauvericin and enniatin B were cytotoxic on these cells. IC50 were equal to 1.0 μM, 2.9 μM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service