Skip to Content
Merck
All Photos(2)

Key Documents

36133

Supelco

Isazophos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17ClN3O3PS
CAS Number:
Molecular Weight:
313.74
Beilstein:
8145294
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1nc(Cl)n(n1)C(C)C

InChI

1S/C9H17ClN3O3PS/c1-5-14-17(18,15-6-2)16-9-11-8(10)13(12-9)7(3)4/h7H,5-6H2,1-4H3

InChI key

XRHGWAGWAHHFLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isazophos may be used as an analytical reference standard for the determination of the analyte in bovine tissues, rice paddies, fruits and vegetables, and tea by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue method for the rapid determination of pesticide residues in tea using ultra performance liquid chromatography Orbitrap high resolution mass spectrometry and in-syringe dispersive solid phase extraction.
Chen H, et al.
ACS Omega, 2(9) (2017)
Needling the pain and comforting the brain: acupuncture in the treatment of chronic migraine.
Shuu-Jiun Wang et al.
Cephalalgia : an international journal of headache, 31(15), 1507-1509 (2011-10-14)
R A Chapman et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 17(4), 355-361 (1982-01-01)
Isofenphos (Amaze (R), Oftanol (R), BAY 92114, 1-methylethyl 2- [[ethoxy [(1-methylethyl)amino] phosphinothioyl] oxy] benzoate) (20% granular) and isazophos (Miral (R), CGA 12223, 0,0-diethyl 0- [1-isopropyl-5-chloro-1,2,4-triazolyl(3)] phosphorothioate) (50% emulsifiable concentrate) were applied at 3.4 Kg AI/ha and incorporated into sand and
C H Sierra-Torres et al.
Mutation research, 415(1-2), 59-67 (1998-08-26)
Miral 500 CS (CAS# 42509-80-8), an organophosphorus insecticide, has been widely used in Columbia to fumigate coffee plantations. Therefore, there is extensive human exposure to this pesticide. Miral's mutagenic and genotoxic activities, however, are not known. In this study, such
N F Motuzinskiĭ et al.
Gigiena i sanitariia, (3)(3), 26-28 (1991-03-01)
On the basis of results of studies on experimental substantiation of MAC for miral in soil it has been found out, that the sub-threshold miral amounts according to the water-migrational, air-migrational, translocational and general sanitary indices of harmfulness equal 0.03

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service