Skip to Content
Merck
All Photos(1)

Key Documents

34108

Supelco

Tributyl phosphate

analytical standard

Synonym(s):

TBP, TBPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3(CH2)3O)3PO
CAS Number:
Molecular Weight:
266.31
Beilstein:
1710584
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

9.2 (vs air)

vapor pressure

27 mmHg ( 178 °C)
7.3 mmHg ( 150 °C)

autoignition temp.

770 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.424 (lit.)

bp

180-183 °C/22 mmHg (lit.)

mp

−79 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCOP(=O)(OCCCC)OCCCC

InChI

1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3

InChI key

STCOOQWBFONSKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tributyl phosphate is an organophosphorus compound, which is widely used as a flame retardant and plasticizer in a variety of industrial products.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tributyl phosphate may be used as an analytical reference standard in the determination of the analyte in:
  • indoor air samples using charcoal tube sampling followed by gas chromatography with mass spectrometry and flame photometric detection
  • human plasma samples using solid-phase microextraction followed by gas chromatography coupled to inductively coupled plasma mass spectrometry

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Analysis of organic esters of plasticizer in indoor air by GC- MS and GC- FPD.
Otake T, et al.
Environmental Science & Technology, 35(15), 3099-3102 (2001)
Monika Shah et al.
Journal of chromatography. A, 1103(2), 329-336 (2005-12-13)
A simple and sensitive method for determination of phosphoric acid triesters at trace levels in human plasma sample is described. In this work, solid-phase microextraction (SPME) is employed as a sample preparation procedure for extraction and pre-concentration of alkyl and
C W Van Neste et al.
Analytical chemistry, 81(5), 1952-1956 (2009-02-04)
Despite its immediate applications, selective detection of trace quantities of surface adsorbed chemicals, such as explosives, without physically collecting the sample molecules is a challenging task. Standoff spectroscopic techniques offer an ideal method of detecting chemicals without using a sample
Muniasamy Neerathilingam et al.
Toxicology letters, 199(1), 10-16 (2010-08-07)
Tributyl phosphate (TBP) is a toxic organophosphorous compound widely used in many industrial applications, including significant usage in nuclear processing. The industrial application of this chemical is responsible for occupational exposure and environmental pollution. In this study, (1)H NMR-based metabonomics
C R Deeken et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 96(2), 199-206 (2011-01-07)
Extracellular matrix (ECM) materials are currently utilized for soft tissue repair applications such as vascular grafts, tendon reconstruction, and hernia repair. These materials are derived from tissues such as human dermis and porcine small intestine submucosa, which must be rendered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service