Skip to Content
Merck
All Photos(1)

Key Documents

19566

Supelco

2,4,6-Triiodophenol

certified reference material, TraceCERT®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I3C6H2OH
CAS Number:
Molecular Weight:
471.80
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

157-159 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Oc1c(I)cc(I)cc1I

InChI

1S/C6H3I3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H

InChI key

VAPDZNUFNKUROY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matilde Parreño et al.
Molecular cancer therapeutics, 5(5), 1166-1175 (2006-05-30)
2,4,6-Triiodophenol (Bobel-24, AM-24) was originally described as a nonsteroid antiinflammatory molecule. We have synthesized three derivatives of Bobel-24 (Bobel-4, Bobel-16, and Bobel-30) and tested their activities as putative antileukemic agents. We have found that Bobel-24 and Bobel-16 were dual inhibitors
José Antonio Castro-Hermida et al.
Veterinary parasitology, 155(3-4), 308-313 (2008-06-27)
The efficacy of the anti-inflammatory drug Bobel-24 against experimental infection by Cryptosporidium parvum was evaluated in neonatal lambs. The animals were treated by oral administration of the drug at 50 or 500 mg/kg of body weight. The prophylactic/therapeutic treatment was
L Xun et al.
Biochemical and biophysical research communications, 174(1), 43-48 (1991-01-15)
Pentachlorophenol (PCP) degrading Flavobacterium sp. ATCC 39723 was found to degrade other polyhalogenated phenolic compounds, including triiodophenol, tribromophenol, and trichlorophenol. Each compound was able to induce the degradation of the other compounds. A PCP Flavobacterium sp. mutant, F-2, was unable
Iñaki F Trocóniz et al.
Pharmaceutical research, 23(7), 1533-1542 (2006-06-20)
The aim of this study was to explore the possibility of achieving a practical dosing regimen for 2,4,6-triiodophenol (AM-24), a new leukotriene B4 (LTB4) synthesis inhibitor. First, a model capable of dealing with the nonlinearity in its pharmacokinetic profile was
L García-Capdevila et al.
Journal of chromatography. B, Biomedical sciences and applications, 708(1-2), 169-175 (1998-07-08)
A rapid and simple HPLC method is described for the determination of Bobel-24 (2,4,6-triiodophenol) and other iodinated derivatives in biological samples. The sample preparation was liquid-liquid extraction before injection onto the HPLC system. 2,6-Diiodo-4-methylphenol was used as internal standard. Separation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service