Skip to Content
Merck
All Photos(1)

Key Documents

05283

Supelco

3-Deoxy-2-keto-6-phosphogluconic acid lithium salt

analytical standard

Synonym(s):

(4S,5R)-4,5-Dihydroxy-2-oxo-6-phosphonooxyhexanoic acid lithium salt, 2-Dehydro-3-deoxy-6-phospho-D-gluconate lithium salt, 2-Keto-3-deoxy-6-phosphogluconate lithium salt, 3-Deoxy-2-keto-D-gluconic acid 6-phosphate lithium salt, 3-Deoxy-D-erythro-hexulosonic acid 6-phosphate lithium salt, KDPG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11O9P · xLi+
CAS Number:
Molecular Weight:
258.12 (free acid basis)
UNSPSC Code:
41116107
NACRES:
NB.21

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

optical activity

[α]/D 10.0±2.0°, 3 hr, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

−20°C

InChI

1S/C6H11O9P/c7-3(1-4(8)6(10)11)5(9)2-15-16(12,13)14/h3,5,7,9H,1-2H2,(H,10,11)(H2,12,13,14)/t3-,5+/m0/s1

InChI key

OVPRPPOVAXRCED-WVZVXSGGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Deoxy-2-keto-6-phosphogluconic acid (KDPG) is a primary metabolite of the Entner-Doudoroff (ED) pathway. It plays an important role in cyanobacteria, algae, higher plants, etc and is estimated to be utilized by 27% of the heterotrophic prokaryotic microorganisms for sugar and sugar acid degradation. Its production using the Entner-Doudoroff dehydratase (EDD) from Caulobacter crescentus (CcEDD) is investigated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic Synthesis of 2-Keto-3-Deoxy-6-Phosphogluconate by the 6-Phosphogluconate-Dehydratase From Caulobacter crescentus
Krevet S, et al.
Frontiers in Bioengineering and Biotechnology, 8, 185-185 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service