01987
N,N-Dimethylallylamine
analytical standard
Synonym(s):
3-Dimethylamino-1-propene, N-Allyl-dimethylamin, DMAA
About This Item
grade
analytical standard
Quality Level
Assay
≥99.0% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤2.0% water
refractive index
n20/D 1.399-1.401
density
0.719 g/mL at 20 °C (lit.)
application(s)
environmental
format
neat
SMILES string
CN(C)CC=C
InChI
1S/C5H11N/c1-4-5-6(2)3/h4H,1,5H2,2-3H3
InChI key
GBCKRQRXNXQQPW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Recommended products
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-4.0 °F - closed cup
Flash Point(C)
-20 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service