Skip to Content
Merck
All Photos(1)

Key Documents

ST1506

Sigma-Aldrich

Anti-SAA1 Mouse mAb (3C11-2C1)

liquid, clone 3C11-2C1, Calbiochem®

Synonym(s):

Anti-Serum Amyloid A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3C11-2C1, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SAA1(6288)

General description

Purified mouse monoclonal antibody. Recognizes the ~15 kDa SAA1 protein.
Recognizes the ~15 kDa SAA1 protein in human spleen and colon adenocarcinoma tissue.
This Anti-SAA1 Mouse mAb (3C11-2C1) is validated for use in Immunoblotting, Paraffin Sections for the detection of SAA1.

Immunogen

Full-length, recombinant human SAA1 (aa 1-123), expressed as a GST fusion protein
NA

Application



Immunoblotting (1-5 g/ml)
Paraffin Sections (1-10 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Human spleen, Human colon adenocarcinoma tissue

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno B Lemire et al.
Journal of applied physiology (Bethesda, Md. : 1985), 113(1), 159-166 (2012-04-21)
Muscle atrophy in chronic obstructive pulmonary disease (COPD) is associated with reduced exercise tolerance, muscle strength, and survival. The molecular mechanisms leading to muscle atrophy in COPD remain elusive. The mitogen-activated protein kinases (MAPKs) such as p38 MAPK and ERK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service