Skip to Content
Merck
All Photos(1)

Key Documents

1.03768

Supelco

2,6-Dimethylphenol

GR for analysis

Synonym(s):

2,6-Dimethylphenol, 2,6-DMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2,6-(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
MDL number:
UNSPSC Code:
12352104
EC Index Number:
209-400-1

vapor pressure

0.2 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

555 °C

potency

296 mg/kg LD50, oral (Rat)
1000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4 % (v/v)

pH

6-7 (20 °C, 8 g/L in H2O)

bp

203 °C

mp

42-46 °C

transition temp

flash point 73 °C

solubility

8 g/L

density

1.03 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/C8H10O/c1-6-4-3-5-7(2)8(6)9/h3-5,9H,1-2H3

InChI key

NXXYKOUNUYWIHA-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 99 % (a/a)
Identity (IR-spectrum): passes test
Melting point: 45 - 48 °C
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Sulfated ash: ≤ 0.05 %
Water (according to Karl Fischer): ≤ 0.1 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service