Key Documents
TEM-CF200-AU-ET
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
Extra Thick, size 200 mesh, box of 50
About This Item
UNSPSC Code:
41111761
size:
200 mesh
packaging:
box of 50
packaging
box of 50
size
200 mesh
General description
A 20-30 nm thick film of pure carbon supported standard squre mesh grids. Gold grid, 200 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)