O0130
Oxalic acid dilithium salt
Synonym(s):
Ethanedioic acid
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C2O4Li2
CAS Number:
Molecular Weight:
101.90
EC Number:
MDL number:
UNSPSC Code:
12352107
eCl@ss:
39021701
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
SMILES string
[Li+].[Li+].[O-]C(=O)C([O-])=O
InChI
1S/C2H2O4.2Li/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2
InChI key
YNQRWVCLAIUHHI-UHFFFAOYSA-L
Application
Oxalic acid dilithium salt may be used to prepare lithium tetrafluorooxalatophosphate, an electrolyte for Li-ion batteries with better thermal stability.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Investigation of lithium tetrafluorooxalatophosphate as a lithium-ion battery electrolyte.
Xu M, et al.
Electrochemical and Solid-State Letters, 12(8), A155-A158 (2009)
Novel Electrolyte for Lithium Ion Batteries: Lithum Tetrafluorooxalatophosphate (LiPF4C2O4).
Xu M, et al.
ECS Transactions, 16(35), 3-11 (2009)
Peter Visser et al.
Faraday discussions, 180, 511-526 (2015-05-01)
Lithium carbonate and lithium oxalate were incorporated as leachable corrosion inhibitors in model organic coatings for the protection of AA2024-T3. The coated samples were artificially damaged with a scribe. It was found that the lithium-salts are able to leach from
Sindhu Jagadamma et al.
PloS one, 7(11), e50434-e50434 (2012-12-05)
Physico-chemical sorption onto soil minerals is one of the major processes of dissolved organic carbon (OC) stabilization in deeper soils. The interaction of DOC on soil solids is related to the reactivity of soil minerals, the chemistry of sorbate functional
Jiří Jan et al.
Water research, 47(2), 547-557 (2012-12-12)
Correct identification of P forms together with their main Fe and Al binding partners in non-calcareous sediments is of crucial importance for evaluation of P cycling in water bodies. In this paper, we assess extraction methods frequently used for this
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service