I15602
Isobutyronitrile
99%
Synonym(s):
2-Methylpropanenitrile, 2-Methylpropionitrile, Isopropyl cyanide
About This Item
Recommended Products
vapor density
2.38 (vs air)
Quality Level
vapor pressure
100 mmHg ( 54.4 °C)
Assay
99%
form
liquid
autoignition temp.
899 °F
refractive index
n20/D 1.372 (lit.)
bp
107-108 °C (lit.)
mp
−72 °C (lit.)
density
0.770 g/mL at 20 °C (lit.)
SMILES string
CC(C)C#N
InChI
1S/C4H7N/c1-4(2)3-5/h4H,1-2H3
InChI key
LRDFRRGEGBBSRN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
46.4 °F - closed cup
Flash Point(C)
8 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service