Skip to Content
Merck
All Photos(1)

Key Documents

900199

Sigma-Aldrich

Iron oxide (II,III), nanoparticles

15 nm avg. part. size (TEM), amine functionalized, 1 mg/mL in H2O

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303

form

dispersion
nanoparticles

Quality Level

concentration

1 mg/mL in H2O

avg. part. size

15 nm (TEM)

functional group

amine

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Iron oxide nanoparticles exhibit biocompatibility and non-toxicity and are used as magnetic resonance imaging contrast agents, gene carriers for gene therapy, therapeutic agents for hyperthermia-based cancer treatment, magnetic sensing probes for in vivo diagnostics and for target specific drug delivery.

Legal Information

Product of Ocean Nanotech, LLC.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianglong Hu et al.
Dalton transactions (Cambridge, England : 2003), 44(9), 3904-3922 (2015-01-13)
Responsive polymeric assemblies and hybrid superstructures fabricated from stimuli-sensitive polymers and inorganic nanoparticles (NPs) have been the subject of extensive investigations during the past few decades due to their distinct advantages such as an improved water solubility, stimuli-responsiveness, excellent biocompatibility
Jin Xie et al.
Theranostics, 2, 122-124 (2012-01-31)
This theme issue provides a timely collection of studies on magnetic nanoparticle-based imaging, bio-sensing, therapy and/or their combinations.
Joan Estelrich et al.
International journal of molecular sciences, 16(4), 8070-8101 (2015-04-14)
In this review, we discuss the recent advances in and problems with the use of magnetically-guided and magnetically-responsive nanoparticles in drug delivery and magnetofection. In magnetically-guided nanoparticles, a constant external magnetic field is used to transport magnetic nanoparticles loaded with

Articles

Professor Hui Mao explores the use of superparamagnetic iron oxide nanoparticles (INOPs) that offer an alternate contrast-enhancing mechanism.

Professor Yadong Yin (University of California Riverside, USA) examines both direct (thermal decomposition, solvothermal, hydrothermal) and indirect (templated) synthesis methods of magnetite nanocrystals and reviews in detail the landscape of these various synthetic methods for magnetite nanocrystal and their applications in magnetic assembly, magnetic hyperthermia, and Li-Ion batteries.

Global Trade Item Number

SKUGTIN
900199-10ML

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service