900093
Iron oxide (II,III), nanoparticles
20 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
About This Item
Recommended Products
form
dispersion
nanoparticles
Quality Level
concentration
1 mg/mL in H2O
avg. part. size
20 nm (TEM)
functional group
streptavidin
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Other Notes
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Professor Hui Mao explores the use of superparamagnetic iron oxide nanoparticles (INOPs) that offer an alternate contrast-enhancing mechanism.
Professor Yadong Yin (University of California Riverside, USA) examines both direct (thermal decomposition, solvothermal, hydrothermal) and indirect (templated) synthesis methods of magnetite nanocrystals and reviews in detail the landscape of these various synthetic methods for magnetite nanocrystal and their applications in magnetic assembly, magnetic hyperthermia, and Li-Ion batteries.
Global Trade Item Number
SKU | GTIN |
---|---|
900093-1ML |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service